Learn More
Perkin Elmer US LLC SeaSpray Glass Nebulizer, 2.0 mL/min, for ELAN/NexION 300/350
Catalog No. 509043893
Encompass_Preferred
The SeaSpray Concentric Nebulizer is ideal for samples with high dissolved solids and offers outstanding nebulization effciency for trace level analysis. It offers freedom from clogging while nebulizing solutions near the limit of solubility of most mineral salts, and conferring signifcant sensitivity gains. All SeaSpray Nebulizers are supplied with the EzyLok argon connector.
Catalog No. 50-904-3893 Supplier Perkin Elmer US LLC Supplier No. N0775340
The SeaSpray Concentric Nebulizer is ideal for samples with high dissolved solids and offers outstanding nebulization effciency for trace level analysis. It offers freedom from clogging while nebulizing solutions near the limit of solubility of most mineral salts, and conferring signifcant sensitivity gains. All SeaSpray Nebulizers are supplied with the EzyLok argon connector.