Learn More
Glass Expansion SeaSpray U-Series Nebulizer 2mL/min (1300017241, 33010017, N0775340, 8003-0490)
SDP
Catalog No. 501907561
Encompass_Preferred
SeaSpray Nebulizer 2.0 mL/min, includes UniFit Connector with 0.75 mm ID x 1.6 mm OD x 1500 mm long tubing (1 pc.) and EzyLok Connector Kit (1 pc.).Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 2.0 mL/min. Recommended not to exceed optimum 2.0 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 1.0 L/min gas flow rate. Tolerates up to 20% TDS. We recommend the use of the Elegra Argon Humidifier to achieve best performance with high TDS.Replacement parts include P/Ns NFTS-16-75 and 502702994.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) SeaSpray to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7561 Supplier Glass Expansion Supplier No. ARG1USS2S
SeaSpray Nebulizer 2.0 mL/min, includes UniFit Connector with 0.75 mm ID x 1.6 mm OD x 1500 mm long tubing (1 pc.) and EzyLok Connector Kit (1 pc.).Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 2.0 mL/min. Recommended not to exceed optimum 2.0 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 1.0 L/min gas flow rate. Tolerates up to 20% TDS. We recommend the use of the Elegra Argon Humidifier to achieve best performance with high TDS.Replacement parts include P/Ns NFTS-16-75 and 502702994.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) SeaSpray to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001