Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sebacic acid

Catalog No. 283258250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-325-8250G 250 g
28-325-85G 5 g
28-325-81KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-325-8250G Supplier Sigma Aldrich Supplier No. 283258250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sebacic acid

Specifications

CAS 111-20-6
Melting Point 133°C to 137°C (lit.)
Boiling Point 294.5°C (100 mmHg)
Molecular Formula C10H18O4
Linear Formula HO2C(CH2)8 CO2H
MDL Number MFCD00004440
Quantity 250 g
Synonym Decanedioic acid
Molecular Weight (g/mol) 202.25
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.