Learn More
Medchemexpress LLC Sebacic acid bis(N-succinimidyl) ester | 23024-29-5 | 97.0% | 25 G
SDP
Catalog No. 5000233925
Encompass_Preferred
Disuccinimidyl sebacate (DSSeb) is a homobifunctional N-hydroxysuccinimide (NHS) ester cross-linker that reacts with primary amines to form stable amide bonds, enabling intermolecular peptide and protein cross-linking and bioconjugation.
  • Homobifunctional NHS-ester for amine coupling.
  • Produces stable amide bonds suitable for peptide and protein cross-linking.
  • Useful for intermolecular cross-linking, bioconjugation, and linker synthesis.
  • Available in multiple packaging sizes, including 25 G.
  • High purity (≈97.0%) suitable for research applications.
Catalog No. 50-002-33925 Supplier Medchemexpress LLC Supplier No. HY11952325G
Disuccinimidyl sebacate (DSSeb) is a homobifunctional N-hydroxysuccinimide (NHS) ester cross-linker that reacts with primary amines to form stable amide bonds, enabling intermolecular peptide and protein cross-linking and bioconjugation.
  • Homobifunctional NHS-ester for amine coupling.
  • Produces stable amide bonds suitable for peptide and protein cross-linking.
  • Useful for intermolecular cross-linking, bioconjugation, and linker synthesis.
  • Available in multiple packaging sizes, including 25 G.
  • High purity (≈97.0%) suitable for research applications.