Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sebacic Acid Pract., 90%, Thermo Scientific™

Catalog No. AC419242500
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419242500 250 g
AC419240010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC419242500 Supplier Thermo Scientific Chemicals Supplier No. 419242500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-20-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
InChI Key CXMXRPHRNRROMY-UHFFFAOYSA-N
Synonym sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
PubChem CID 5192
ChEBI CHEBI:41865
IUPAC Name decanedioic acid
SMILES C(CCCCC(=O)O)CCCC(=O)O

Specifications

Color White
Density 1.21
Infrared Spectrum Authentic
Linear Formula HOCO(CH2)8COOH
Quantity 250 g
Beilstein 02, IV, 2078
Merck Index 15, 8552
Formula Weight 202.25
Percent Purity ≥88%
Physical Form Crystalline Powder
Chemical Name or Material Sebacic acid, pract., 90%
Show More Show Less
EINECSNumber 203-845-5
RTECSNumber VS0875000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.