Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sebacoyl chloride

Catalog No. 13178450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-178-450G 50 g
13-178-4250G 250 g
13-178-4500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-178-450G Supplier Sigma Aldrich Supplier No. 13178450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sebacoyl chloride

Specifications

CAS 111-19-3
Melting Point -5°C to +3°C
Density 1.121 g/mL (at 25°C)
Boiling Point 168°C (2 mmHg)
Molecular Formula C10H16Cl2O2
Refractive Index n20/D 1.468 (literature)
Linear Formula ClCO(CH2)8 COCl
Quantity 50 g
MDL Number MFCD00000770
Synonym Sebacyl chloride
Molecular Weight (g/mol) 239.14
Percent Purity 92%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.