Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sebacoyl Chloride/Hexane Solution, 4% v/v
SCI_ED

Catalog No. S25517A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25517A 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25517A

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Nylon production

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 111-19-3 , 110-54-3
Molecular Formula C10H16Cl2O2
Molecular Weight (g/mol) 239.136
InChI Key WMPOZLHMGVKUEJ-UHFFFAOYSA-N
Synonym sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
PubChem CID 66072
IUPAC Name decanedioyl dichloride
SMILES C(CCCCC(=O)Cl)CCCC(=O)Cl

Specifications

Assay Percent Range 96% v/v
Density 0.70g/mL at 20°C
Packaging 4%
Quantity 100 mL
Chemical Name or Material Sebacoyl Chloride/Hexane Solution, 4% v/v
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.