Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sebacoyl-L-Carnitine Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022427 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2427 50 mg
11-102-2426 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2427 Supplier MilliporeSigma Supelco Supplier No. 1632950MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 102636-85-1 (Free Acid)
Molecular Formula C17H31NO6 · xLi+
Linear Formula C17H31NO6 · xLi+
Quantity 50 mg
Formula Weight 345.43 (Free Acid Basis)
Percent Purity ≥93% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.