Learn More
Cole-Parmer Thermoplastic (SEBS) Tubing, 3/16" ID x 5/16" OD; 50 Ft
Supplier:  Cole-Parmer 5011975
Encompass_Preferred
Designed for the most demanding biopharmaceutical applications, the thermoplastic SEBS (styrene-ethylene-butylene-styrene) tubing exhibits superior gas barrier properties and excellent pressure performance. Use the tubing in peristaltic pumps, single-use assemblies, process filling, and sampling devices.
Thermoplastic (SEBS) tubing does not contain BPA, phthalates, PVC, or peroxides that can leach out into the fluid, making it ideal for pharmaceutical applications, buffer and media preparation, and cell media processing. For an added level of particle control, a liner is placed between the tubing package and the box.
Catalog No. 50-256-3450