Learn More
NANOMICRO TECHNOLOGIES INC BioCore SEC-120 LC Analytical Column, 3um, 4.6x300 mm, Stainless Steel
Catalog No. NC2815331
Encompass_Preferred
BioCore SEC is a family of high performance, size exclusion chromatography columns designed for separating a broad range of biomolecules based on size. This column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, high-strength, monodispersed silica particles, combined with well-established column packing processes. BioCore SEC-120 columns are suited for aggregate determination of peptides, small proteins and related substances.
Catalog No. NC2815331 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B21303001204630S
Add to Cart
Add to Cart
BioCore SEC is a family of high performance, size exclusion chromatography columns designed for separating a broad range of biomolecules based on size. This column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, high-strength, monodispersed silica particles, combined with well-established column packing processes. BioCore SEC-120 columns are suited for aggregate determination of peptides, small proteins and related substances.