Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Protein BEH SEC Column, 125Å, 2.5 μm, 4.6 mm x 150 mm

Catalog No. NC1767262 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge Protein BEH SEC Column separates proteins from 1,000 to 80,000 Daltons using 125Å pore, 2.5 μm Ethylene Bridged Hybrid (BEH) particles with a diol-bonded surface. Rigorous QC ensures consistent performance, pH stability, efficiency, peak form, and capacity, making it ideal for LC-MS assays and quality control.

  • Separates proteins from 1,000 to 80,000 Daltons.
  • Features 125Å pore size and 2.5 μm BEH particles.
  • Diol-bonded surface coating for enhanced performance.
  • Ensures excellent pH stability, efficiency, and peak form.
  • Compatible with UHPLC and HPLC systems.
  • Operating pH range: 2.5 to 8.
  • Max operating pressure: 5500 psi (379 Bar).
  • Max operating temperature: 60°C.
  • Dimensions: 4.6 mm ID x 150 mm length.

Catalog No. NC1767262 Supplier Waters Corp Supplier No. 186009171
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge Protein BEH SEC Column separates proteins from 1,000 to 80,000 Daltons using 125Å pore, 2.5 μm Ethylene Bridged Hybrid (BEH) particles with a diol-bonded surface. Rigorous QC ensures consistent performance, pH stability, efficiency, peak form, and capacity, making it ideal for LC-MS assays and quality control.

  • Separates proteins from 1,000 to 80,000 Daltons.
  • Features 125Å pore size and 2.5 μm BEH particles.
  • Diol-bonded surface coating for enhanced performance.
  • Ensures excellent pH stability, efficiency, and peak form.
  • Compatible with UHPLC and HPLC systems.
  • Operating pH range: 2.5 to 8.
  • Max operating pressure: 5500 psi (379 Bar).
  • Max operating temperature: 60°C.
  • Dimensions: 4.6 mm ID x 150 mm length.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.