Learn More
NANOMICRO TECHNOLOGIES INC BioCore SEC-300 LC Analytical Column, 5um, 7.8x300 mm, Stainless Steel
Catalog No. NC2815250
Encompass_Preferred
BioCore SEC is a family of high performance, size exclusion chromatography columns designed for separating a broad range of biomolecules based on size. The column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, high-strength, monodispersed silica particles, combined with well-established column packing processes. BioCore SEC-300 columns are suited for aggregate and fragment determination in antibodies and related substances.
Catalog No. NC2815250 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B21305003007830S
Add to Cart
Add to Cart
BioCore SEC is a family of high performance, size exclusion chromatography columns designed for separating a broad range of biomolecules based on size. The column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, high-strength, monodispersed silica particles, combined with well-established column packing processes. BioCore SEC-300 columns are suited for aggregate and fragment determination in antibodies and related substances.