Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl Acetoacetate 98.0+%, TCI America™
SDP

Catalog No. A081525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A081525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A081525ML Supplier TCI America Supplier No. A081525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13562-76-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00059392
InChI Key QSTNBMLCULGCQE-UHFFFAOYSA-N
Synonym Acetoacetic Acid sec-Butyl Ester
PubChem CID 139498
IUPAC Name butan-2-yl 3-oxobutanoate
SMILES CCC(C)OC(=O)CC(=O)C

Specifications

Color Colorless
Boiling Point 194°C
Quantity 25 mL
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Acetoacetate
EINECSNumber (2)-1475&(2)-1505
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.