Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sec-Butyl Alcohol, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S000551 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000551 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: Flammable liquid and vapour, causes serious eye irritation, may cause respiratory irritation, may cause drowsiness or dizziness.
Catalog No. S000551 Supplier Aldon Corporation Supplier No. IS11080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning: Flammable liquid and vapour, causes serious eye irritation, may cause respiratory irritation, may cause drowsiness or dizziness.

Chemical Identifiers

Molecular Weight (g/mol) 74.12 g/mol

Specifications

Quantity 500 mL
Boiling Point 97°C to 99°C
Percent Purity 100%
Grade Laboratory
Chemical Name or Material Sec-Butyle Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.