Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sec-Butyllithium Solution

Catalog No. 195596800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
4 x 25 mL
800 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-559-6800ML 800 mL
19-559-6100ML 100 mL
19-559-625ML 4 x 25 mL
19-559-650ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 19-559-6800ML Supplier Sigma Aldrich Supplier No. 195596800ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

sec-Butyllithium solution (1.4M in cyclohexane) has been used in the multi-step synthesis of 5-methyl-5,6-dihydrothymidine (5-MDHT) from thymidine. It has also been used in the copper-catalyzed asymmetric allylic alkylation (AAA) of allyl bromides, chlorides, and ethers in the presence of a chiral ligand.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 598-30-1
Density 0.769 g/mL at 25°C
Molecular Formula C4H9Li
Linear Formula CH3CH2CH(CH3)Li
MDL Number MFCD00009323
Quantity 800 mL
Synonym Lithium-2-butanide; s-BuLi
Molecular Weight (g/mol) 64.06 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.