Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sec-Butylmagnesium Chloride Solution

Catalog No. 224421800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-442-1800ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 22-442-1800ML Supplier Sigma Aldrich Supplier No. 224421800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

sec-Butylmagnesium chloride solution is a Grignard reagent, it can be used in: Metal catalyzed cross-coupling reactions with non-activated aryl chlorides.The synthesis of various alkoxytrichlorosilanes by reacting with tetrachlorosilane and alcohols.

Specifications

CAS 1306-38-3
Density 7.13 g/mL at 25°C
Molecular Formula CeO2
Linear Formula CeO2
MDL Number MFCD00010933
Quantity 100 g
Molecular Weight (g/mol) 172.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.