Learn More
Azzota Corp SEC-C 0.5mm electro cell
Catalog No. 5000887154
Encompass_Preferred
SEC-C Thin Layer Quartz Crystal Spectroelectrochemical cell Pathlength 0 5mm Quartz Glass (190nm-2500nm) Dimensions 12 5mmx12 5mmx42mm (Height)
Catalog No. 50-008-87154 Supplier Azzota Corp Supplier No. SECC05
SEC-C Thin Layer Quartz Crystal Spectroelectrochemical cell Pathlength 0 5mm Quartz Glass (190nm-2500nm) Dimensions 12 5mmx12 5mmx42mm (Height)