Learn More
Azzota Corp SEC-C 1.0mm electro cell
Catalog No. 5000887042
Encompass_Preferred
SEC-C Thin Layer Quartz Crystal Spectroelectrochemical cell Pathlength 1 0mm Quartz Glass (190nm-2500nm) Dimensions 12 5mmx12 5mmx42mm (Height)
Catalog No. 50-008-87042 Supplier Azzota Corp Supplier No. SECC10
SEC-C Thin Layer Quartz Crystal Spectroelectrochemical cell Pathlength 1 0mm Quartz Glass (190nm-2500nm) Dimensions 12 5mmx12 5mmx42mm (Height)