Learn More
Azzota Corp SEC-C 2.0mm electro cell
Catalog No. 5000887172
Encompass_Preferred
SEC-C Thin Layer Quartz Crystal Spectroelectrochemical cell Pathlength 2 0mm Quartz Glass (190nm-2500nm) Dimensions 12 5mmx12 5mmx42mm (Height)
Catalog No. 50-008-87172 Supplier Azzota Corp Supplier No. SECC20
SEC-C Thin Layer Quartz Crystal Spectroelectrochemical cell Pathlength 2 0mm Quartz Glass (190nm-2500nm) Dimensions 12 5mmx12 5mmx42mm (Height)