Learn More
Waters Corp SEC Guard Column 5µm 500Å 4.6mm ID
Supplier:  Waters Corp WTC050N5G
Encompass_Preferred
SEC Guard Column 5µm 500Å 4.6mm ID. Appropriate for SEC-MALS of adeno-associated virus (AAV) samples.
Catalog No. NC1793708