Learn More
YMC America, Inc SEC MAB 250A 3um 250x.3mm 1/16
SDP
Catalog No. 5000879313
Encompass
YMC-SEC MAB is a silica based size exclusion chromatography column specifically designed for antibody separations Optimized base material design and surface modification provide superior separation of antibody monomers and their aggregates/fragments with very low nonspecific adsorption This is ideal for the quality control assessment of antibody drugs YMC-SEC MAB exhibits excellent resolution and peak shape over a wide molecular weight range of 10 kDa to 700 kDa
Catalog No. 50-008-79313 Supplier YMC America, Inc Supplier No. DLM25S0325H0AU
YMC-SEC MAB is a silica based size exclusion chromatography column specifically designed for antibody separations Optimized base material design and surface modification provide superior separation of antibody monomers and their aggregates/fragments with very low nonspecific adsorption This is ideal for the quality control assessment of antibody drugs YMC-SEC MAB exhibits excellent resolution and peak shape over a wide molecular weight range of 10 kDa to 700 kDa