Learn More
Aviva Systems Biology Corp SEC14L2 Antibody - middle region (ARP31825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEC14L2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SEC14L2. Alternate names: SPF, TAP, TAP1, C22orf6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54330