Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603426 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-426 50 kg Poly Bottle
18-603-420 25 g Poly Bottle
18-603-421 100 g Poly Bottle
18-603-422 500 g Poly Bottle
18-603-423 1 kg Poly Bottle
18-603-424 5 kg Poly Bottle
18-603-425 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-603-426 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IM10550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IM105, 288-32-4, C3H4N2

Spectrum™ Chemical Imidazole, Reagent is a solid that has no color and is water soluble. It is classified as an diazole and an alkaloid and is an aromatic heterocycle. It belongs to the class of antifungals known as azole and is also used as a corrosion inhibitor on certain metals, notably copper. Imidazole is incorporated into many biological molecules, primarily histidine. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

CAS Max % 100%
Boiling Point 256°C (492.8°F)
Packaging Poly Bottle
Quantity 50 kg
UN Number UN3263
Percent Purity 99%
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.