Learn More
Aviva Systems Biology Corp Sec14l3 Antibody - C-terminal region (ARP55733_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55733_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sec14l3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Spf2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-072