Learn More
Aviva Systems Biology Corp SEC23B Antibody - middle region (ARP52162_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52162_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEC23B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SEC23B. Alternate names: CDAII, CDAN2, CDA-II, HEMPASApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60690