Learn More
Aviva Systems Biology Corp Sec31a Antibody - N-terminal region (ARP55111_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55111_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sec31a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: VAP1, Sec31l1, MGC93320, Sec31aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-006