Learn More
Medchemexpress LLC Sec61-IN-1 (compound A317) | 2484865-42-9 | 99.7% | 430.53 g/mol | C23H22N6OS | 5 MG
SDP
Catalog No. 5000221153
Encompass_Preferred
Sec61-IN-1 (compound A317) is a small-molecule inhibitor of the Sec61 translocon used as a research tool to study protein translocation and Sec61-mediated pathways. Manufacturer documentation reports molecular formula C23H22N6OS, molecular weight 430.53 g/mol, and purity of 99.7%. The compound has been reported to target glioma cells and to enhance T cell cytotoxic effects in experimental studies.
  • High purity (99.7%) suitable for research use.
  • Selective inhibition of Sec61-mediated protein translocation.
  • Reported activity against glioma cells and enhancement of T cell cytotoxicity.
  • Supplied as a five mg vial for small-scale in vitro or in vivo preparation.
  • Includes supporting documentation: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-21153 Supplier Medchemexpress LLC Supplier No. HY1396155MG
Sec61-IN-1 (compound A317) is a small-molecule inhibitor of the Sec61 translocon used as a research tool to study protein translocation and Sec61-mediated pathways. Manufacturer documentation reports molecular formula C23H22N6OS, molecular weight 430.53 g/mol, and purity of 99.7%. The compound has been reported to target glioma cells and to enhance T cell cytotoxic effects in experimental studies.
  • High purity (99.7%) suitable for research use.
  • Selective inhibition of Sec61-mediated protein translocation.
  • Reported activity against glioma cells and enhancement of T cell cytotoxicity.
  • Supplied as a five mg vial for small-scale in vitro or in vivo preparation.
  • Includes supporting documentation: data sheet, certificate of analysis, and safety data sheet.