Learn More
Medchemexpress LLC Sec61-IN-1 (compound A317) | 2484865-42-9 | 99.7% | C23H22N6OS | 10MG
SDP
Catalog No. 5000259603
Encompass_Preferred
Sec61-IN-1 (compound A317) is a small-molecule inhibitor of the Sec61 translocon developed for research use. It selectively inhibits Sec61-mediated protein translocation, shows cytotoxic activity in certain glioma models, and can enhance T cell-mediated cytotoxicity in vitro; it also shows measurable brain exposure in mouse studies. Supplied as a solid or as a DMSO solution for laboratory studies.
  • Potent Sec61 inhibition in vitro.
  • Enhances T cell-mediated cytotoxicity against glioma cells.
  • Demonstrated activity in SEC61G-EGFR amplified glioblastoma models.
  • Detectable brain exposure after intravenous dosing in mice.
  • Available as solid and as a DMSO solution for formulation flexibility.
  • High purity suitable for research (99.7%).
  • Recommended storage conditions provided to preserve stability.
Catalog No. 50-002-59603 Supplier Medchemexpress LLC Supplier No. HY13961510MG
Sec61-IN-1 (compound A317) is a small-molecule inhibitor of the Sec61 translocon developed for research use. It selectively inhibits Sec61-mediated protein translocation, shows cytotoxic activity in certain glioma models, and can enhance T cell-mediated cytotoxicity in vitro; it also shows measurable brain exposure in mouse studies. Supplied as a solid or as a DMSO solution for laboratory studies.
  • Potent Sec61 inhibition in vitro.
  • Enhances T cell-mediated cytotoxicity against glioma cells.
  • Demonstrated activity in SEC61G-EGFR amplified glioblastoma models.
  • Detectable brain exposure after intravenous dosing in mice.
  • Available as solid and as a DMSO solution for formulation flexibility.
  • High purity suitable for research (99.7%).
  • Recommended storage conditions provided to preserve stability.