Learn More
ABclonal Technology SEC62 Rabbit mAb
SDP
Catalog No. 5000006952
Encompass_Preferred
The Sec61 complex is the central component of the protein translocation apparatus of the endoplasmic reticulum (ER) membrane The protein encoded by this gene and SEC63 protein are found to be associated with ribosome-free SEC61 complex It is speculated that Sec61-Sec62-Sec63 may perform post-translational protein translocation into the ER The Sec61-Sec62-Sec63 complex might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway The encoded protein is an integral membrane protein located in the rough ER
Catalog No. 50-000-06952 Supplier ABclonal Technology Supplier No. A2400120UL
The Sec61 complex is the central component of the protein translocation apparatus of the endoplasmic reticulum (ER) membrane The protein encoded by this gene and SEC63 protein are found to be associated with ribosome-free SEC61 complex It is speculated that Sec61-Sec62-Sec63 may perform post-translational protein translocation into the ER The Sec61-Sec62-Sec63 complex might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway The encoded protein is an integral membrane protein located in the rough ER