Learn More
Seca Corp 201 Retractable waist circumference measuring tape, cm-only, 0 - 205 centimeters
SDP
Catalog No. NC2176222 Shop All Seca Corp Products
Encompass_Preferred
Today’s circumference tapes also have to satisfy high medical demands. That is why seca has developed the seca 201 which can measure girth with millimeter precision. The high-quality mechanics guarantee that the 205-centimeters long tape unwinds easily and locks in place precisely. The housing sits comfortably and securely in the user’s hand and is tough enough to withstand accidental dropping.• Retractable measuring tape for precise body measurements in centimeters • Printed scale on side makes results easy-to-read • Compact and easy to store • Measuring range: 0 – 205 cm• Graduation: 1 mm
This item is not returnable. View return policy
Catalog No. NC2176222 Supplier Seca Corp Supplier No. 2011717009
Today’s circumference tapes also have to satisfy high medical demands. That is why seca has developed the seca 201 which can measure girth with millimeter precision. The high-quality mechanics guarantee that the 205-centimeters long tape unwinds easily and locks in place precisely. The housing sits comfortably and securely in the user’s hand and is tough enough to withstand accidental dropping.• Retractable measuring tape for precise body measurements in centimeters • Printed scale on side makes results easy-to-read • Compact and easy to store • Measuring range: 0 – 205 cm• Graduation: 1 mm