Learn More
Research Products International Corp Secador Desiccator Cabinet, Vertical, Clear, 1.2 Cubic Feet, 13 1/4 x 17 1/4 x 12 1/2 Inches
SDP
Catalog No. 501977095
Encompass
The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products. Choose from an extensive selection of sizes, styles and colors of desiccator cabinets to meet your specific dry storage needs. U.S. Patents: 6,834,920; 7,318,630 and D474,626.
•    Polycarbonate construction features a UV light blocker and is resistant to staining, crazing and chemical attack.
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7095 Supplier Research Products International Corp Supplier No. 420112
The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products. Choose from an extensive selection of sizes, styles and colors of desiccator cabinets to meet your specific dry storage needs. U.S. Patents: 6,834,920; 7,318,630 and D474,626.
•    Polycarbonate construction features a UV light blocker and is resistant to staining, crazing and chemical attack.
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.