Learn More
Medchemexpress LLC HY-100559 5mg Medchemexpress, SecinH3 CAS:853625-60-2 Purity:>98%
SDP
Catalog No. 501931881
Encompass_Preferred
Medchemexpress, HY-100559 5mg SecinH3 CAS:853625-60-2 SecinH3 is an antagonist of cytohesins with IC 50 s of 5.4 μM, 2.4 μM, 5.4 μM, 5.6 μM, 5.6 μM and 65 μM for hCyh1 , hCyh2 , mCyh3 , hCyh3 , drosophila steppke and yGea2-S7 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1881 Supplier Medchemexpress LLC Supplier No. HY1005595MG
Medchemexpress, HY-100559 5mg SecinH3 CAS:853625-60-2 SecinH3 is an antagonist of cytohesins with IC 50 s of 5.4 μM, 2.4 μM, 5.4 μM, 5.6 μM, 5.6 μM and 65 μM for hCyh1 , hCyh2 , mCyh3 , hCyh3 , drosophila steppke and yGea2-S7 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.