Learn More
Proteintech Group Inc SECISBP2 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 127981AP150UL
Encompass_Preferred
The SECISBP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SECISBP2. This antibody recognizes human, mouse, rat antigen. The SECISBP2 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Catalog No. 50-225-6382