Learn More
Medchemexpress LLC HY-103713 10mg , Seclidemstat CAS:1423715-37-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10371310MG
Encompass_Preferred
HY-103713 10mg Seclidemstat CAS: 1423715-37-0 Seclidemstat is a potent noncompetitive and reversible KDM1A (LSD1) inhibitor (Ki=31 nM, IC50=13 nM). Seclidemstat promotes antitumor immunity in switch/sucrose nonfermentable (SWI/SNF) complex mutated ovarian cancer, as well as inhibit virus production, viral DNA replication, and late gene expression. Seclidemstat can be used for the research of Ewing Sarcoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0454