Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Seclidemstat (mesylate) | 2044953-70-8 | 99.8% | 547.04 | C21H27ClN4O7S2 | 50 MG
SDP

Catalog No. 5000243453
Encompass_Preferred

Seclidemstat mesylate is the mesylate salt of a reversible, non-covalent small-molecule inhibitor of lysine-specific demethylase 1 (LSD1) used in preclinical oncology and immuno-oncology research. It is supplied as a solid research reagent with supporting safety and quality documentation for laboratory use.

  • LSD1 inhibitor for cancer and immuno-oncology research.
  • Mesylate salt form for improved stability and handling.
  • High purity (>99.8%) suitable for analytical and biological assays.
  • Molecular weight 547.04 and formula C21H27ClN4O7S2.
  • Available in small research pack sizes, including 50 mg.
  • Includes SDS and datasheet for handling and quality information.

Catalog No. 50-002-43453 Supplier Medchemexpress LLC Supplier No. HY103713A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seclidemstat mesylate is the mesylate salt of a reversible, non-covalent small-molecule inhibitor of lysine-specific demethylase 1 (LSD1) used in preclinical oncology and immuno-oncology research. It is supplied as a solid research reagent with supporting safety and quality documentation for laboratory use.

  • LSD1 inhibitor for cancer and immuno-oncology research.
  • Mesylate salt form for improved stability and handling.
  • High purity (>99.8%) suitable for analytical and biological assays.
  • Molecular weight 547.04 and formula C21H27ClN4O7S2.
  • Available in small research pack sizes, including 50 mg.
  • Includes SDS and datasheet for handling and quality information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.