Learn More
Medchemexpress LLC Seclidemstat mesylate | 2044953-70-8 | 99.8% | 547.04 g/mol | C20H23ClN4O4S·CH4O3S | 100 MG
SDP
Catalog No. 5000243848
Encompass_Preferred
Seclidemstat mesylate is the mesylate salt of seclidemstat (SP-2577), a reversible, noncompetitive inhibitor of lysine-specific demethylase 1 (KDM1A/LSD1) used in preclinical research. It exhibits potent biochemical activity and is investigated for antitumor immunity and cancer research applications, including studies of Ewing sarcoma and SWI/SNF-mutated ovarian cancer.
  • Reversible, noncompetitive KDM1A (LSD1) inhibitor.
  • Demonstrates potent biochemical activity (Ki ≈ 31 nM; IC50 ≈ 13 nM).
  • High purity suitable for research (99.8%).
  • Provided as a mesylate salt to improve handling and stability.
  • Supplied as a powder; store at 4°C away from moisture and light.
  • Used in oncology and virology research applications.
Catalog No. 50-002-43848 Supplier Medchemexpress LLC Supplier No. HY103713A100MG
Seclidemstat mesylate is the mesylate salt of seclidemstat (SP-2577), a reversible, noncompetitive inhibitor of lysine-specific demethylase 1 (KDM1A/LSD1) used in preclinical research. It exhibits potent biochemical activity and is investigated for antitumor immunity and cancer research applications, including studies of Ewing sarcoma and SWI/SNF-mutated ovarian cancer.
  • Reversible, noncompetitive KDM1A (LSD1) inhibitor.
  • Demonstrates potent biochemical activity (Ki ≈ 31 nM; IC50 ≈ 13 nM).
  • High purity suitable for research (99.8%).
  • Provided as a mesylate salt to improve handling and stability.
  • Supplied as a powder; store at 4°C away from moisture and light.
  • Used in oncology and virology research applications.