Learn More
Medchemexpress LLC Seclidemstat mesylate | 2044953-70-8 | 99.8% | 547.04 g·mol⁻¹ | C21H27ClN4O7S2 | 10 MG
SDP
Catalog No. 5000241886
Encompass_Preferred
Seclidemstat mesylate is a research-grade, reversible, noncompetitive inhibitor of lysine-specific demethylase 1 (KDM1A/LSD1) used in preclinical studies. It demonstrates potent biochemical activity and is applied in oncology and virology research, including investigations of SWI/SNF-complex-mutated tumors and fusion-positive sarcomas.
  • Reversible, noncompetitive KDM1A (LSD1) inhibitor suitable for biochemical and cell-based assays.
  • Demonstrated potency with Ki = 31 nM and IC50 = 13 nM.
  • High-purity solid form for reliable experimental reproducibility.
  • Available in multiple packaging sizes, including small research quantities.
  • Stable when stored under recommended conditions for both short- and long-term use.
Catalog No. 50-002-41886 Supplier Medchemexpress LLC Supplier No. HY103713A10MG
Seclidemstat mesylate is a research-grade, reversible, noncompetitive inhibitor of lysine-specific demethylase 1 (KDM1A/LSD1) used in preclinical studies. It demonstrates potent biochemical activity and is applied in oncology and virology research, including investigations of SWI/SNF-complex-mutated tumors and fusion-positive sarcomas.
  • Reversible, noncompetitive KDM1A (LSD1) inhibitor suitable for biochemical and cell-based assays.
  • Demonstrated potency with Ki = 31 nM and IC50 = 13 nM.
  • High-purity solid form for reliable experimental reproducibility.
  • Available in multiple packaging sizes, including small research quantities.
  • Stable when stored under recommended conditions for both short- and long-term use.