Learn More
Medchemexpress LLC Seclidemstat mesylate | 2044953-70-8 | 99.8% | 547.04 g·mol⁻¹ | C21H27ClN4O7S2 | 25 MG
SDP
Catalog No. 5000243618
Encompass_Preferred
Seclidemstat mesylate is a reversible, noncompetitive small-molecule inhibitor of lysine-specific demethylase 1 (LSD1/KDM1A) supplied as the mesylate salt for preclinical research. It is provided as a high-purity solid and formulated options for biochemical and cell-based studies in oncology and antiviral research.
  • Reversible, noncompetitive LSD1 inhibitor (reported Ki = 31 nM, IC50 = 13 nM).
  • High purity (99.8%) suitable for biochemical and cell-based assays.
  • Available in multiple quantities, including 25 MG and 10 mM solution in DMSO.
  • Mesylate salt form for improved handling and stability.
  • Applicable to research in Ewing sarcoma, SWI/SNF-mutated cancers, and antiviral studies.
Catalog No. 50-002-43618 Supplier Medchemexpress LLC Supplier No. HY103713A25MG
Seclidemstat mesylate is a reversible, noncompetitive small-molecule inhibitor of lysine-specific demethylase 1 (LSD1/KDM1A) supplied as the mesylate salt for preclinical research. It is provided as a high-purity solid and formulated options for biochemical and cell-based studies in oncology and antiviral research.
  • Reversible, noncompetitive LSD1 inhibitor (reported Ki = 31 nM, IC50 = 13 nM).
  • High purity (99.8%) suitable for biochemical and cell-based assays.
  • Available in multiple quantities, including 25 MG and 10 mM solution in DMSO.
  • Mesylate salt form for improved handling and stability.
  • Applicable to research in Ewing sarcoma, SWI/SNF-mutated cancers, and antiviral studies.