Learn More
MSC Guardair Second Stage Venturi Shut-Off Valve, Automatic Performance Control
Catalog No. 502836517
Encompass
The Second Stage Venturi Shut-Off Valve is designed to automatically control vacuum performance, delivering high vacuum flow and high vacuum head as needed. It is an integral component of PowerQUAD vacuum generating engines, specifically managing the second venturi stage.
  • Automatically controls vacuum performance
  • Delivers high vacuum flow when required
  • Delivers high vacuum head when required
  • Automatically closes off the second stage venturi based on vacuuming demand
  • Requires periodic checking to ensure free operation
  • Replace if the valve does not move freely, or is damaged, ripped, or torn
Catalog No. 50-283-6517 Supplier MSC Supplier No. 64361686
The Second Stage Venturi Shut-Off Valve is designed to automatically control vacuum performance, delivering high vacuum flow and high vacuum head as needed. It is an integral component of PowerQUAD vacuum generating engines, specifically managing the second venturi stage.
  • Automatically controls vacuum performance
  • Delivers high vacuum flow when required
  • Delivers high vacuum head when required
  • Automatically closes off the second stage venturi based on vacuuming demand
  • Requires periodic checking to ensure free operation
  • Replace if the valve does not move freely, or is damaged, ripped, or torn