Learn More
Apexbio Technology LLC Secnidazole 3366-95-8 10mg
SDP
Catalog No. 502240725
Encompass_Preferred
Secnidazole (CAS 3366-95-8) is a small-molecule inhibitor targeting bacterial quorum sensing (QS) pathways It is designed to disrupt QS-regulated virulence processes thereby attenuating pathogenic potential independently of bacterial growth Secnidazole exerts its biological activity primarily through inhibition of QS-mediated mechanisms particularly in Pseudomonas aeruginosa In in vitro studies Secnidazole demonstrates inhibitory activity with an IC50 value of approximately 0 25 M in Pseudomonas aeruginosa quorum-sensing inhibition assays Based on these pharmacological properties Secnidazole holds research potential in investigations of virulence factor modulation bacterial infections such as bacterial vaginosis and parasitic protozoal infections including amoebiasis and giardiasis
This item is not returnable. View return policy
Catalog No. 50-224-0725 Supplier Apexbio Technology LLC Supplier No. B216210
Secnidazole (CAS 3366-95-8) is a small-molecule inhibitor targeting bacterial quorum sensing (QS) pathways It is designed to disrupt QS-regulated virulence processes thereby attenuating pathogenic potential independently of bacterial growth Secnidazole exerts its biological activity primarily through inhibition of QS-mediated mechanisms particularly in Pseudomonas aeruginosa In in vitro studies Secnidazole demonstrates inhibitory activity with an IC50 value of approximately 0 25 M in Pseudomonas aeruginosa quorum-sensing inhibition assays Based on these pharmacological properties Secnidazole holds research potential in investigations of virulence factor modulation bacterial infections such as bacterial vaginosis and parasitic protozoal infections including amoebiasis and giardiasis