Learn More
Cayman Chemical Secnidazole, Purity: ≥98%, CAS: 3366-95-8, 10 mg
SDP
Catalog No. 502985106
Encompass_Preferred
A 5-nitroimidazole derivative with antiparasitic and antibiotic activity; exhibits a mean minimum inhibitory concentration of 2.59 µM against 16 clinical isolates of B. fragilis
Catalog No. 50-298-5106 Supplier Cayman Chemical Supplier No. 2123610 MG
A 5-nitroimidazole derivative with antiparasitic and antibiotic activity; exhibits a mean minimum inhibitory concentration of 2.59 µM against 16 clinical isolates of B. fragilis