Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Seco-DUBA | 1227961-59-2 | 526.97 | C29H23ClN4O4 | 10 MG
SDP

Catalog No. 5000203563
Encompass_Preferred

Seco-DUBA is a duocarmycin (DUBA) prodrug intended for use as an antibody-drug conjugate (ADC) payload. The molecule contains two hydroxyl groups that enable conjugation to antibodies via suitable linkers, and it is supplied in small research quantities for ADC development and synthetic work.

  • Duocarmycin prodrug suitable for ADC payloads.
  • Two hydroxyl groups for linker coupling.
  • Intended for use in antibody-drug conjugate synthesis and research.
  • Chemical formula: C29H23ClN4O4.
  • Molecular weight: 526.97.
  • Available in small package sizes for research (for example, 10 mg).

Catalog No. 50-002-03563 Supplier Medchemexpress LLC Supplier No. HY132180A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seco-DUBA is a duocarmycin (DUBA) prodrug intended for use as an antibody-drug conjugate (ADC) payload. The molecule contains two hydroxyl groups that enable conjugation to antibodies via suitable linkers, and it is supplied in small research quantities for ADC development and synthetic work.

  • Duocarmycin prodrug suitable for ADC payloads.
  • Two hydroxyl groups for linker coupling.
  • Intended for use in antibody-drug conjugate synthesis and research.
  • Chemical formula: C29H23ClN4O4.
  • Molecular weight: 526.97.
  • Available in small package sizes for research (for example, 10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.