Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Seco-DUBA | 1227961-59-2 | MFCD06642603 | 526.97 g/mol | C29H23ClN4O4 | 5 MG
SDP

Catalog No. 5000203122
Encompass_Preferred

Seco-DUBA is a duocarmycin (DUBA) prodrug used as an antibody-drug conjugate (ADC) payload precursor for research and development. It contains two hydroxyl groups that enable coupling to an antibody via a linker, and it functions as a DNA-alkylating cytotoxin for targeted delivery in ADC synthesis.

  • Contains two hydroxyl groups for antibody conjugation.
  • Designed as an ADC payload precursor for conjugation chemistry.
  • Reported molecular formula C29H23ClN4O4 and molecular weight 526.97 g/mol.
  • Available in small research pack sizes suitable for R&D use.

Catalog No. 50-002-03122 Supplier Medchemexpress LLC Supplier No. HY132180A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seco-DUBA is a duocarmycin (DUBA) prodrug used as an antibody-drug conjugate (ADC) payload precursor for research and development. It contains two hydroxyl groups that enable coupling to an antibody via a linker, and it functions as a DNA-alkylating cytotoxin for targeted delivery in ADC synthesis.

  • Contains two hydroxyl groups for antibody conjugation.
  • Designed as an ADC payload precursor for conjugation chemistry.
  • Reported molecular formula C29H23ClN4O4 and molecular weight 526.97 g/mol.
  • Available in small research pack sizes suitable for R&D use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.