Learn More
Medchemexpress LLC Seco rapamycin ethyl ester | 00-00-0 | >98.0% | 942.23 g/mol | C53H83NO13 | 5 MG
SDP
Catalog No. 5000263186
Encompass_Preferred
Seco rapamycin ethyl ester is the ethyl-ester open-ring metabolite of rapamycin supplied for research use. It is reported not to affect mTOR function and is used in biochemical and pharmacological studies of rapamycin metabolism and structure-activity relationships. Molecular formula C53H83NO13; molecular weight 942.23 g/mol.
  • Open-ring rapamycin metabolite for research applications.
  • Reportedly does not affect mTOR function, useful for mechanistic studies.
  • Suitable for biochemical and pharmacological assays.
  • Available in small research quantities for compound screening.
  • Supplied as a characterized compound; verify batch certificate for exact purity.
Catalog No. 50-002-63186 Supplier Medchemexpress LLC Supplier No. HY1337705MG
Seco rapamycin ethyl ester is the ethyl-ester open-ring metabolite of rapamycin supplied for research use. It is reported not to affect mTOR function and is used in biochemical and pharmacological studies of rapamycin metabolism and structure-activity relationships. Molecular formula C53H83NO13; molecular weight 942.23 g/mol.
  • Open-ring rapamycin metabolite for research applications.
  • Reportedly does not affect mTOR function, useful for mechanistic studies.
  • Suitable for biochemical and pharmacological assays.
  • Available in small research quantities for compound screening.
  • Supplied as a characterized compound; verify batch certificate for exact purity.