Learn More
Medchemexpress LLC Seco Rapamycin sodium salt | 148554-65-8 | 90.6% | 936.15 | 25 MG
SDP
Catalog No. 5000402432
Encompass_Preferred
Seco Rapamycin sodium salt is the ring-opened product of Rapamycin. It is reported not to affect the mTOR function.
  • Ring-opened product of Rapamycin
  • Reported not to affect mTOR function
  • Available in various quantities
  • For research use only
Catalog No. 50-004-02432 Supplier Medchemexpress LLC Supplier No. 25MGHY19555A
Seco Rapamycin sodium salt is the ring-opened product of Rapamycin. It is reported not to affect the mTOR function.
  • Ring-opened product of Rapamycin
  • Reported not to affect mTOR function
  • Available in various quantities
  • For research use only