Learn More
Medchemexpress LLC Seco rapamycin sodium salt | 148554-65-8 | 90.6% | 936.15 g/mol | C51H78NNaO13 | 10 MG
SDP
Catalog No. 5000294629
Encompass_Preferred
Seco Rapamycin sodium salt is the ring-opened derivative of rapamycin supplied as the monosodium salt. It is reported not to affect mTOR function and has been characterized in vitro for NADPH-dependent conversion to dihydro sirolimus (M2) in human tissue homogenates and Caco-2 models. Supplied as a white to yellow solid with specified purity and recommended low-temperature storage.
  • Ring-opened derivative of rapamycin
  • Reported not to affect mTOR function
  • Metabolized in vitro to dihydro sirolimus (M2) in an NADPH-dependent manner
  • Provided as a monosodium solid salt
  • Recommended storage -20°C; protect from light and store under nitrogen
  • Suitable for in vitro research and metabolic studies
Catalog No. 50-002-94629 Supplier Medchemexpress LLC Supplier No. HY19555A10MG
Seco Rapamycin sodium salt is the ring-opened derivative of rapamycin supplied as the monosodium salt. It is reported not to affect mTOR function and has been characterized in vitro for NADPH-dependent conversion to dihydro sirolimus (M2) in human tissue homogenates and Caco-2 models. Supplied as a white to yellow solid with specified purity and recommended low-temperature storage.
  • Ring-opened derivative of rapamycin
  • Reported not to affect mTOR function
  • Metabolized in vitro to dihydro sirolimus (M2) in an NADPH-dependent manner
  • Provided as a monosodium solid salt
  • Recommended storage -20°C; protect from light and store under nitrogen
  • Suitable for in vitro research and metabolic studies