Learn More
Cayman Chemical Seco RapamycInsodIum salt 10mg
SDP
Catalog No. 502971708
Encompass_Preferred
A nonenzyme-dependent degradation product of rapamycin resulting from ester hydration followed by dehydration; secreted from cells by P-glycoprotein; poorly activates mTOR but, like rapamycin, inhibits the proteasome
Catalog No. 50-297-1708 Supplier Cayman Chemical Supplier No. 1615810 MG
A nonenzyme-dependent degradation product of rapamycin resulting from ester hydration followed by dehydration; secreted from cells by P-glycoprotein; poorly activates mTOR but, like rapamycin, inhibits the proteasome