Learn More
SECO™ Rhino Shield 502 Barrier Bags
Catalog No. 19045802 Shop All Seco Industries Products
Click to view available options
Quantity:
100
Catalog No. Quantity
19-045-802 100
1 options
Catalog No. 19-045-802 Supplier SECO™ Supplier No. 5022224CL
Ideal for applications requiring a flexible, heat—sealable barrier material
  • Material: Metalized polyester, polyethylene, aluminum foil and anti-static polyethylene
  • May be used to protect extremely critical or sensitive devices
  • 0.0036 mil (0.00014 in.) thickness
  • May be used with devices susceptible to damage by E.S.D, E.M.I, moisture or corrosion
  • Available with a press to close zipper, slide rite zipper, or open top
  • Color: Opaque
  • Custom sizes available
Compliance
Meets military specifications of Mil-Prf-81705C Ty-1, CL-1 (mil spec printed)
Specifications
Closure Type Press to close zipper, slide rite zipper or open top
Color Silver
Material Metalized Polyester, Polyethylene, Aluminum Foil, Anti-static Polyethylene
Quantity 100