Learn More
Cayman Chemical -SecoIsolarIcIresInol 10mg
SDP
Catalog No. 502948443
Encompass_Preferred
A lignan with diverse biological activities; inhibits α-glucosidase and α-amylase in a cell-free assay at 20 µM; reduces adiponectin production by, and triglyceride accumulation in, 3T3-L1 adipocytes at 0.3 mM; stimulates IgM production in HB4C5 hybridoma cells from 0.1-100 µM
Catalog No. 50-294-8443 Supplier Cayman Chemical Supplier No. 3315510 MG
A lignan with diverse biological activities; inhibits α-glucosidase and α-amylase in a cell-free assay at 20 µM; reduces adiponectin production by, and triglyceride accumulation in, 3T3-L1 adipocytes at 0.3 mM; stimulates IgM production in HB4C5 hybridoma cells from 0.1-100 µM