Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Secoisolariciresinol | 145265-02-7 | 5 MG
SDP

Catalog No. 5000419016
Encompass_Preferred

(+)-Secoisolariciresinol is a lignan compound and a (+)-enantiomer of Secoisolariciresinol. It is intended for research use only.

  • Purity of 99.66%
  • Molecular weight: 362.42
  • Chemical formula: C20H26O6
  • Appearance: solid
  • Color: white to off-white
  • Soluble in DMSO (requires sonication)
  • Compatible with various in vivo dissolution protocols

Catalog No. 50-004-19016 Supplier Medchemexpress LLC Supplier No. HYN81725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Secoisolariciresinol is a lignan compound and a (+)-enantiomer of Secoisolariciresinol. It is intended for research use only.

  • Purity of 99.66%
  • Molecular weight: 362.42
  • Chemical formula: C20H26O6
  • Appearance: solid
  • Color: white to off-white
  • Soluble in DMSO (requires sonication)
  • Compatible with various in vivo dissolution protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.